siRNA Details
Gene Symbol: 9830107B12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177896.1 | NP_001171367.1 | 2 | 520 | ||
NM_001177897.1 | NP_001171368.1 | 2 | 373 | |||
NM_177824.4 | NP_808492.1 | 2 | 520 | |||
GenBank mRNA | AK079398.1 | - | 2 | 481 | ||
AK137568.1 | - | 2 | 318 | |||
EF646801.1 | - | 221 |
