siRNA Details
Gene Symbol: Gm815 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033407.2 | NP_001028579.1 | 3, 4 | 268 | ||
GenBank mRNA | AK136142.1 | - | 3, 4 | 269 | ||
BC147433.1 | - | 3, 4 | 236 | 9056486 | ||
BC172070.1 | - | 3, 4 | 269 |
