siRNA Details
Gene Symbol: Dennd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166501.1 | NP_001159973.1 | 6, 7 | 536 | ||
NM_181347.3 | NP_851992.1 | 5, 6 | 402 | |||
GenBank mRNA | AK089338.1 | - | 1 | 230 | ||
AK155727.1 | - | 5, 6 | 403 | |||
BC042698.1 | - | 6, 7 | 459 | |||
BC141388.1 | - | 5, 6 | 398 | 9056001 | ||
BC141389.1 | - | 5, 6 | 398 | 9056002 |
