siRNA Details
Gene Symbol: Rnf224 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033410.2 | NP_001028582.1 | 2 | 310 | ||
GenBank mRNA | AK146725.1 | - | 2 | 311 | ||
BC147425.1 | - | 1 | 503 | 9056501 | ||
BC147426.1 | - | 1 | 503 | 9056480 |
