siRNA Details
Gene Symbol: Adamts3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081401.2 | NP_001074870.1 | 18, 19 | 3032 | ||
NM_177872.2 | NP_808540.2 | 18, 19 | 3035 | |||
GenBank mRNA | BC158091.1 | - | 18, 19 | 2636 | 9088176 | |
BC158105.1 | - | 18, 19 | 2633 | 9088181 |
