siRNA Details
Gene Symbol: Kctd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159941.1 | NP_001153413.1 | 6, 7 | 805 | ||
NM_026145.4 | NP_080421.2 | 6, 7 | 802 | |||
GenBank mRNA | AK028657.1 | - | 6, 7 | 769 | ||
AK041516.1 | - | 6, 7 | 769 | |||
AK145295.1 | - | 6, 7 | 781 | |||
AK147714.1 | - | 6, 7 | 747 | |||
AK161199.1 | - | 6, 7 | 759 | |||
AK163351.1 | - | 6, 7 | 756 | |||
AK166505.1 | - | 6, 7 | 781 | |||
BC006935.1 | - | 6, 7 | 783 | 3601917 |
