siRNA Details
Gene Symbol: Gm765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128092.1 | NP_001121564.1 | 3 | 310 | ||
GenBank mRNA | BC147266.1 | - | 3 | 310 | 9056347 | |
BC147267.1 | - | 3 | 310 | 9056335 |
Gene Symbol: Gm32592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK139454.1 | - | 1 | 1338 |
