siRNA Details
Gene Symbol: Trim67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198632.2 | NP_941034.2 | 10 | 3162 | ||
GenBank mRNA | BC094596.1 | - | 10 | 3089 | 5705515 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trim67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198632.2 | NP_941034.2 | 10 | 3162 | ||
GenBank mRNA | BC094596.1 | - | 10 | 3089 | 5705515 |