siRNA Details
Gene Symbol: AI593442 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286641.1 | NP_001273570.1 | 2 | 587 | ||
NM_178906.5 | NP_849237.2 | 2 | 538 | |||
GenBank mRNA | AK033979.1 | - | 2 | 537 | ||
BC109328.2 | - | 2 | 407 | 40051267 | ||
BM933125.1 | - | 1 | 289 |
