siRNA Details
Gene Symbol: Slc9a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177909.5 | NP_808577.3 | 16 | 1923 | ||
GenBank mRNA | AK037058.1 | - | 16 | 1907 | ||
AK156494.1 | - | 16 | 1884 | |||
AK172687.1 | - | 16 | 1895 | |||
BC099942.1 | - | 16 | 1897 | 30103308 | ||
BC153825.1 | - | 16 | 1901 | 40093280 |
