siRNA Details
Gene Symbol: Fnd3c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033424.3 | NP_001028596.2 | 10 | 1558 | ||
GenBank mRNA | AK030291.1 | - | 10 | 1563 | ||
AK146031.1 | - | 10 | 1559 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fnd3c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033424.3 | NP_001028596.2 | 10 | 1558 | ||
GenBank mRNA | AK030291.1 | - | 10 | 1563 | ||
AK146031.1 | - | 10 | 1559 |