siRNA Details
Gene Symbol: G630025P09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014547.1 | - | 1257 | |||
AK036110.1 | - | 1129 | ||||
AK137322.1 | - | 1 | 1492 | |||
AK139243.1 | - | 1 | 1482 | |||
AK165723.1 | - | 1 | 1492 |
Gene Symbol: Tmem102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033433.4 | NP_001028605.2 | 1 | 462 | ||
GenBank mRNA | AK143262.1 | - | 1 | 462 |
