siRNA Details
Gene Symbol: Gm5141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256065.1 | NP_001242994.1 | 4 | 1622 | ||
GenBank mRNA | BC064714.1 | - | 4 | 1622 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm5141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256065.1 | NP_001242994.1 | 4 | 1622 | ||
GenBank mRNA | BC064714.1 | - | 4 | 1622 |