siRNA Details
Gene Symbol: Dgkh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081336.1 | NP_001074805.1 | 22 | 2664 | ||
NM_001253766.1 | NP_001240695.1 | 20 | 2352 | |||
NM_001281794.1 | NP_001268723.1 | 20 | 2352 | |||
GenBank mRNA | AK083819.1 | - | 20 | 2353 | ||
AK133314.1 | - | 21 | 3127 | |||
BC117904.1 | - | 20 | 2336 | |||
BC127173.1 | - | 20 | 2336 | 40097093 | ||
CK780920.1 | - | 2 | 187 |
