siRNA Details
Gene Symbol: A630095N17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243090.1 | NP_001230019.1 | 2 | 175 | ||
NM_001243091.1 | NP_001230020.1 | 2 | 88 | |||
GenBank mRNA | AK080399.1 | - | 2 | 89 |
