siRNA Details
Gene Symbol: Kpna7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013774.2 | NP_001013796.2 | 10 | 1508 | ||
GenBank mRNA | AK136027.1 | - | 6 | 1026 | ||
AY950703.1 | - | 10 | 1509 | |||
FJ717332.1 | - | 8 | 1275 | |||
FJ717333.1 | - | 9 | 1338 |
