siRNA Details
Gene Symbol: Scgb2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207262.2 | NP_997145.2 | 2 | 240 | ||
GenBank mRNA | AY370634.1 | - | 2 | 248 | ||
CK616217.1 | - | 2 | 240 | |||
CK616805.1 | - | 284 | ||||
GU269234.1 | - | 2 | 227 |
