siRNA Details
Gene Symbol: Cnep1r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029074.3 | NP_083350.2 | 4 | 278 | ||
GenBank mRNA | AK017198.1 | - | 4 | 239 | ||
AK146578.1 | - | 4 | 269 | |||
BC018498.1 | - | 3 | 176 | |||
BC094385.1 | - | 4 | 237 | |||
BC118981.1 | - | 4 | 239 | |||
BC148291.2 | - | 4 | 239 | 40103934 | ||
BY124150.1 | - | 4 | 280 |
