siRNA Details
Gene Symbol: Cnep1r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029074.3 | NP_083350.2 | 5 | 394 | ||
GenBank mRNA | AK017198.1 | - | 4 | 897 | ||
AK146578.1 | - | 5 | 385 | |||
AK217813.1 | - | 2 | 124 | |||
BC018498.1 | - | 4 | 292 | |||
BC094385.1 | - | 4 | 895 | |||
BC118981.1 | - | 5 | 350 | |||
BC148291.2 | - | 5 | 355 | 40103934 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001106173, XM_008772374.
Gene Ontology Categories:
