siRNA Details
Gene Symbol: Cnep1r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029074.3 | NP_083350.2 | 5, 6 | 422 | ||
GenBank mRNA | AK017198.1 | - | 4, 5 | 925 | ||
AK146578.1 | - | 5, 6 | 413 | |||
AK217813.1 | - | 2, 3 | 152 | |||
BC018498.1 | - | 4, 5 | 320 | |||
BC118981.1 | - | 5, 6 | 378 | |||
BC148291.2 | - | 5, 6 | 383 | 40103934 |
