siRNA Details
Gene Symbol: Gse1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145896.1 | NP_001139368.1 | 5 | 794 | ||
NM_001145897.1 | NP_001139369.1 | 5 | 738 | |||
NM_198671.2 | NP_941073.2 | 5 | 926 | |||
GenBank mRNA | AK033263.1 | - | 5 | 741 | ||
AK042915.1 | - | 5 | 742 | |||
AK122216.1 | - | 2 | 76 | |||
AK154829.1 | - | 5 | 773 | |||
BC043094.1 | - | 5 | 926 | 6405464 |
