siRNA Details
Gene Symbol: AB124611 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198794.1 | NP_001185723.1 | 2 | 424 | ||
NM_206536.2 | NP_996259.2 | 2 | 424 | |||
GenBank mRNA | AB124611.1 | - | 2 | 324 | ||
AB124612.1 | - | 2 | 324 | |||
AK137593.1 | - | 3 | 511 | |||
AK154442.1 | - | 2 | 411 | |||
AK162293.1 | - | 2 | 411 | |||
AK168311.1 | - | 2 | 411 | |||
AK170200.1 | - | 2 | 411 | |||
BU696592.1 | - | 2 | 425 |
