siRNA Details
Gene Symbol: AB124611 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198794.1 | NP_001185723.1 | 7 | 688 | ||
NM_206536.2 | NP_996259.2 | 6 | 598 | |||
GenBank mRNA | AB124611.1 | - | 7 | 588 | ||
AB124612.1 | - | 6 | 498 | |||
AK137593.1 | - | 8 | 775 | |||
AK154442.1 | - | 7 | 675 | |||
AK162293.1 | - | 7 | 675 | |||
AK168311.1 | - | 7 | 675 | |||
AK170200.1 | - | 7 | 675 |
