siRNA Details
Gene Symbol: Cep162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199316.2 | NP_955020.2 | 4 | 555 | ||
GenBank mRNA | AK135482.1 | - | 4 | 556 | ||
AK163175.1 | - | 4 | 541 | |||
BC004073.1 | - | 4 | 314 | |||
BC064713.1 | - | 4 | 311 | |||
BC150998.1 | - | 4 | 360 | 9087911 | ||
BC151006.1 | - | 4 | 360 | 9087919 | ||
BC151104.1 | - | 4 | 360 | 9088017 |
