siRNA Details
Gene Symbol: Fbxw24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013776.4 | NP_001013798.2 | 9 | 1272 | ||
GenBank mRNA | BC080701.1 | - | 9 | 1273 | 30473126 | |
CO808277.1 | - | 4 | 620 | |||
DV651634.1 | - | 5 | 710 | |||
GQ418172.1 | - | 9 | 1273 |
