siRNA Details
Gene Symbol: Kctd16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026135.1 | NP_080411.1 | 2 | 1191 | ||
GenBank mRNA | AK005863.1 | - | 1 | 243 | ||
AK006368.1 | - | 1 | 243 | |||
AK015313.1 | - | 2 | 525 | |||
AK220381.1 | - | 2 | 1165 | |||
BC049734.1 | - | 3 | 459 |
