siRNA Details
Gene Symbol: Kctd16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026135.1 | NP_080411.1 | 1 | 418 | ||
GenBank mRNA | AK220381.1 | - | 1 | 392 |

More Information
Other Species Targeted

Gene Ontology Categories:
