siRNA Details
Gene Symbol: BC031181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001181.3 | NP_001001181.1 | 2 | 317 | ||
GenBank mRNA | AK077642.1 | - | 2 | 250 | ||
AK131658.1 | - | 2 | 314 | |||
AK180483.1 | - | 2 | 222 | |||
AK187367.1 | - | 2 | 223 | |||
BC016084.1 | - | 2 | 317 | 4501468 | ||
BC027819.1 | - | 2 | 331 | 5320851 | ||
BC031181.1 | - | 2 | 312 | 4988367 |
