siRNA Details
Gene Symbol: 4930404N11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014836.3 | NP_001014836.2 | 2 | 241 | ||
GenBank mRNA | AK015088.1 | - | 2 | 212 | ||
BC049656.1 | - | 2 | 211 | |||
BC139256.1 | - | 2 | 237 | 8862278 | ||
BC145589.1 | - | 2 | 237 | |||
BC145960.1 | - | 2 | 237 | 40131263 | ||
BU945663.1 | - | 2 | 244 |
Gene Symbol: Mfsd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK149678.1 | - | 10 | 2527 | ||
AK154386.1 | - | 10 | 2523 | |||
AK154902.1 | - | 10 | 2601 | |||
AK170185.1 | - | 10 | 2541 | |||
AK171084.1 | - | 10 | 2521 |
