siRNA Details
Gene Symbol: Vrtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033776.2 | NP_001028948.1 | 2 | 1250 | ||
NM_001168588.1 | NP_001162060.1 | 3 | 1662 | |||
NM_001168589.2 | NP_001162061.1 | 2 | 1320 | |||
GenBank mRNA | BC103771.1 | - | 2 | 1250 | 30935479 |
