siRNA Details
Gene Symbol: Zfp708 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012325.2 | NP_001012325.2 | 4 | 647 | ||
NM_001012448.2 | NP_001012450.2 | 4 | 559 | |||
NM_001012449.2 | NP_001012451.2 | 3 | 551 | |||
NM_001292066.1 | NP_001278995.1 | 4 | 516 | |||
GenBank mRNA | AK132353.1 | - | 4 | 518 | ||
BB612019.1 | - | 4 | 518 | |||
BC089368.1 | - | 4 | 526 | |||
BG080714.2 | - | 4 | 610 | |||
CF174027.1 | - | 4 | 647 |
