siRNA Details
Gene Symbol: Otulin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013792.2 | NP_001013814.2 | 7 | 1001 | ||
GenBank mRNA | AK134955.1 | - | 7 | 836 | ||
AK150371.1 | - | 7 | 1004 | |||
BC028541.1 | - | 2 | 201 | |||
BC087945.1 | - | 4 | 602 | 30505408 |
