siRNA Details
Gene Symbol: Smim22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253796.1 | NP_001240725.1 | 2, 3 | 250 | ||
NM_001253797.1 | NP_001240726.1 | 2, 3 | 250 | |||
NM_001253802.1 | NP_001240731.1 | 2, 3 | 185 | |||
NM_001253803.1 | NP_001240732.1 | 2, 3 | 185 | |||
GenBank mRNA | AW985536.1 | - | 2, 3 | 176 | ||
BB610432.1 | - | 1, 2 | 87 | |||
BC047216.1 | - | 2, 3 | 229 | |||
CJ043101.1 | - | 2, 3 | 186 | |||
CJ237462.1 | - | 2, 3 | 247 | |||
CJ320043.1 | - | 2, 3 | 247 |
