siRNA Details
Gene Symbol: AA467197 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004174.1 | NP_001004174.1 | 3, 4 | 202 | ||
GenBank mRNA | AK141297.1 | - | 2, 3 | 305 | ||
AK150561.1 | - | 3, 4 | 192 | |||
BC049633.1 | - | 3, 4 | 202 | 6774154 |
