siRNA Details
Gene Symbol: I830077J02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033780.3 | NP_001028952.1 | 5, 6 | 359 | ||
GenBank mRNA | AK152510.1 | - | 5, 6 | 362 | ||
BC147821.1 | - | 5, 6 | 299 | 9087538 | ||
BC147828.1 | - | 5, 6 | 299 | 9087545 |
