siRNA Details
Gene Symbol: Clec2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277202.1 | NP_001264131.1 | 1 | 45 | ||
GenBank mRNA | HQ875060.1 | - | 1 | 23 | ||
JF792240.1 | - | 1 | 45 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Clec2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277202.1 | NP_001264131.1 | 1 | 45 | ||
GenBank mRNA | HQ875060.1 | - | 1 | 23 | ||
JF792240.1 | - | 1 | 45 |