siRNA Details
Gene Symbol: Zdhhc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017968.2 | NP_001017968.2 | 7, 8 | 1072 | ||
GenBank mRNA | AY668950.1 | - | 6, 7 | 651 | ||
BC145714.1 | - | 6, 7 | 703 | |||
BC145716.1 | - | 6, 7 | 703 | |||
DQ113650.1 | - | 1, 2 | 53 |
