siRNA Details
Gene Symbol: Klf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021366.3 | NP_067341.2 | 1 | 385 | ||
GenBank mRNA | AF251796.1 | - | 1 | 384 | ||
AF252285.1 | - | 1 | 363 | |||
AJ245644.1 | - | 1 | 320 | |||
AJ275987.1 | - | 1 | 336 | |||
AK155298.1 | - | 1 | 336 | |||
BC051557.1 | - | 1 | 336 |
