siRNA Details
Gene Symbol: Park2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016694.3 | NP_057903.1 | 7 | 842 | ||
GenBank mRNA | AB019558.1 | - | 7 | 813 | ||
AF250293.1 | - | 7 | 842 | |||
BC113204.1 | - | 7 | 825 | 40047891 | ||
FJ555554.1 | - | 5 | 595 | |||
FJ555555.1 | - | 2 | 253 | |||
FJ555556.1 | - | 6 | 575 | |||
FJ555557.1 | - | 6 | 700 |
