siRNA Details
Gene Symbol: Irx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018885.2 | NP_061373.1 | 3 | 715 | ||
GenBank mRNA | AF124732.1 | - | 3 | 420 | ||
AK132571.1 | - | 3 | 715 | |||
BC126942.1 | - | 3 | 353 | 40089986 |
