siRNA Details
Gene Symbol: Nus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030250.2 | NP_084526.1 | 4 | 981 | ||
GenBank mRNA | AK155620.1 | - | 4 | 983 | ||
AK162260.1 | - | 2 | 143 | |||
AK167741.1 | - | 4 | 970 | |||
AK167939.1 | - | 4 | 974 | |||
BC003223.1 | - | 4 | 940 | 3482163 | ||
BC018372.1 | - | 4 | 956 | 3499868 |
