siRNA Details
Gene Symbol: Tet1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253857.1 | NP_001240786.1 | 11 | 5163 | ||
NM_027384.1 | NP_081660.1 | 10 | 5067 | |||
GenBank mRNA | AI605989.1 | - | 2 | 318 | ||
AK129421.1 | - | 11 | 3416 | |||
GU079948.1 | - | 11 | 5163 |
