siRNA Details
Gene Symbol: Sco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040026.1 | NP_001035115.1 | 3, 4 | 530 | ||
GenBank mRNA | AK011251.1 | - | 3, 4 | 546 | ||
AK017794.1 | - | 3, 4 | 531 | |||
AK139512.1 | - | 3, 4 | 501 | |||
AK167305.1 | - | 3, 4 | 395 | |||
BC139008.1 | - | 3, 4 | 525 | 8862030 | ||
BC139009.1 | - | 3, 4 | 525 | 8862031 |
