siRNA Details
Gene Symbol: Ube2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016786.3 | NP_058066.2 | 3, 4 | 443 | ||
GenBank mRNA | AB011081.1 | - | 3, 4 | 381 | ||
AK018366.1 | - | 3, 4 | 417 | |||
AK035371.1 | - | 3, 4 | 340 | |||
AK036068.1 | - | 3, 4 | 384 | |||
AK048194.1 | - | 3, 4 | 434 | |||
AK139564.1 | - | 3, 4 | 460 | |||
AK160728.1 | - | 3, 4 | 403 | |||
AK162142.1 | - | 3, 4 | 384 | |||
AK166647.1 | - | 3, 4 | 352 | |||
BC002013.1 | - | 3, 4 | 291 | 3496393 | ||
BC085311.1 | - | 3, 4 | 369 | 30148009 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001106006, XM_006250934, FQ233155, FQ234076, FQ226880.
Gene Ontology Categories:
