siRNA Details
Gene Symbol: Reck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016678.2 | NP_057887.2 | 21 | 2897 | ||
GenBank mRNA | AB006960.1 | - | 21 | 2827 | ||
AK042266.1 | - | 22 | 3076 | |||
AK143657.1 | - | 21 | 2876 | |||
AK157885.1 | - | 21 | 2886 | |||
BC053025.1 | - | 21 | 2836 | |||
BC132197.1 | - | 21 | 2925 | 40130474 | ||
BC138065.1 | - | 21 | 2925 | 8861084 | ||
BC145133.1 | - | 21 | 2926 |
