siRNA Details
Gene Symbol: Cntn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017383.3 | NP_059079.2 | 17 | 2453 | ||
GenBank mRNA | AB032602.1 | - | 16 | 2139 | ||
AK052972.1 | - | 18 | 2346 | |||
BC076594.1 | - | 17 | 2453 | 30649661 |
