siRNA Details
Gene Symbol: Rfk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019437.3 | NP_062310.1 | 3, 4 | 614 | ||
GenBank mRNA | AK002806.1 | - | 3, 4 | 575 | ||
AK008352.1 | - | 3, 4 | 544 | |||
AK010607.1 | - | 3, 4 | 543 | |||
BC033521.1 | - | 3, 4 | 516 | 4527980 | ||
BC051021.1 | - | 3, 4 | 525 | 6315425 |
