siRNA Details
Gene Symbol: 1810014B01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007490.1 | - | 2, 3 | 200 | ||
AK012122.1 | - | 2, 3 | 245 | |||
AK017634.1 | - | 255 | ||||
AK019015.1 | - | 2, 3 | 202 | |||
AK049515.1 | - | 1, 2 | 217 | |||
AK090016.1 | - | 2, 3 | 242 |
Gene Symbol: 1190007I07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135567.1 | NP_001129039.1 | 2, 3 | 381 | ||
NM_001135568.1 | NP_001129040.1 | 2, 3 | 252 | |||
NM_001135569.1 | NP_001129041.1 | 2, 3 | 193 | |||
GenBank mRNA | AK004515.1 | - | 2, 3 | 243 | ||
AK045413.1 | - | 2, 3 | 265 | |||
BC038652.1 | - | 2, 3 | 211 | |||
BC065417.1 | - | 2, 3 | 215 | |||
DV649038.1 | - | 2, 3 | 187 |

More Information
Other Species Targeted

Gene Ontology Categories:
