siRNA Details
Gene Symbol: Arhgap27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205236.1 | NP_001192165.1 | 4 | 1246 | ||
NM_133715.5 | NP_598476.2 | 2 | 215 | |||
GenBank mRNA | AK046763.1 | - | 2 | 216 | ||
AK154926.1 | - | 4 | 1247 |
