siRNA Details
Gene Symbol: Wdr70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081402.1 | NP_001074871.1 | 3 | 199 | ||
GenBank mRNA | AK014750.1 | - | 3 | 196 | ||
AK159714.1 | - | 3 | 181 | |||
AK160933.1 | - | 3 | 134 | |||
BC037184.1 | - | 3 | 200 | |||
BC051482.1 | - | 3 | 166 | |||
BC061236.1 | - | 3 | 199 |
Gene Symbol: Gm1943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK170003.1 | - | 1 | 185 | ||
BC024407.1 | - | 1 | 178 |
